Learn More
1-(4-Nitrophenyl)glycerol, 99%
Product Code. 11344356
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344356 1 g 1g
11364356 250 mg 250mg
11354356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11344356 Supplier Thermo Scientific Alfa Aesar Supplier No. L19968.03
 Restricted Item
In Stock
Add to basket
CAS: 2207-68-3 | C9H11NO5 | 213.189 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2207-68-3
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00017037
InChI Key IUZVZBIQZKBWCC-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl propane-1,2,3-triol, p-nitrophenylglycerol, 1-4-nitrophenyl glycerol, 1-4-nitrophenyl-1,2,3-propanetriol, 1,2,3-propanetriol, 1-4-nitrophenyl, 1-4-nitrophenyl glycerol pnpg, 4-nitrophenylglycerol, pubchem9886, para-nitrophenylglycerol, maybridge1_002064
PubChem CID 102764
IUPAC Name 1-(4-nitrophenyl)propane-1,2,3-triol
SMILES C1=CC(=CC=C1C(C(CO)O)O)[N+](=O)[O-]
Specifications
Melting Point 95°C to 97°C
Odor Odorless
Quantity 1 g
Beilstein 7376091
Sensitivity Hygroscopic
Formula Weight 213.19
Percent Purity 99%
Chemical Name or Material 1-(4-Nitrophenyl)glycerol