accessibility menu, dialog, popup

UserName

HOHOOHNOO

CAS RN 2207-68-3

IUPAC Name: 1-(4-nitrophenyl)propane-1,2,3-triol
Synonyms: 1-(4-nitrophenyl)propane-1,2,3-triol
Molecular Weight (g/mol): 213.19
Molecular Formula: C9H11NO5
InChi Key: IUZVZBIQZKBWCC-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)