missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-1-(4-Fluorophenyl)ethylamine, 97%, Thermo Scientific™

Product Code. 11341627
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341627 5 g 5g
11331627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341627 Supplier Alfa Aesar™ Supplier No. L04575.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-40-7
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD00041323
InChI Key QGCLEUGNYRXBMZ-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
PubChem CID 123063
IUPAC Name 1-(4-fluorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)F)N

Specifications

Density 1.03
pH 10
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Odor Amine-like
Refractive Index 1.502
Quantity 5 g
UN Number UN2735
Beilstein 2802417
Sensitivity Air Sensitive
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material (+/-)-1-(4-Fluorophenyl)ethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.