accessibility menu, dialog, popup

UserName

H3C(S)NH3F

CAS RN 403-40-7

IUPAC Name: (1S)-1-(4-fluorophenyl)ethan-1-aminium
Synonyms: (1S)-1-(4-fluorophenyl)ethanaminium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: QGCLEUGNYRXBMZ-LURJTMIESA-O
SMILES: C[C@H]([NH3+])C1=CC=C(F)C=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)