missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-fluorobenzene, 99%, Thermo Scientific™

Product Code. 11449187
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449187 25 g 25g
11439187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449187 Supplier Thermo Scientific Alfa Aesar Supplier No. B21282.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,4-Dichloro-2-fluorobenzene is used as organic synthesis intermediates

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-59-4
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.99
MDL Number MFCD00060656
InChI Key BJQTYCQGIXZSNM-UHFFFAOYSA-N
Synonym 2,5-dichlorofluorobenzene, benzene, 1,4-dichloro-2-fluoro, 1,4-dichloro-2-fluoro-benzene, 2,5-dichloro-1-fluorobenzene, 1,4-dichlor-2-fluorbenzol, 2,5-dichloro-fluorobenzene, pubchem3454, acmc-1ae1b, 2,5-dichloro fluorobenzene, 1-fluor-2,5-dichlorbenzol
PubChem CID 136173
IUPAC Name 1,4-dichloro-2-fluorobenzene
SMILES FC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 4°C
Density 1.383
Boiling Point 168°C
Flash Point 65°C (149°F)
Refractive Index 1.5245
Quantity 25 g
Beilstein 2355590
Solubility Information Difficult to mix.
Formula Weight 165
Percent Purity 99%
Chemical Name or Material 1,4-Dichloro-2-fluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.