accessibility menu, dialog, popup

UserName

FClCl

CAS RN 348-59-4

IUPAC Name: 1,4-dichloro-2-fluorobenzene
Synonyms: 1,4-dichloro-2-fluorobenzene
Molecular Weight (g/mol): 164.99
Molecular Formula: C6H3Cl2F
InChi Key: BJQTYCQGIXZSNM-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results