missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne, 98+%

Product Code. p-7045922
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313186 5 g 5g
11323186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313186 Supplier Thermo Scientific Alfa Aesar Supplier No. L12492.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5768-10-5 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5768-10-5
MDL Number MFCD00021401

Specifications

Melting Point 174°C to 175°C
Molecular Formula C9H8O4
Quantity 5 g
Beilstein 1246867
Synonym myristicin aldehyde, 3-methoxy-4,5-methylenedioxybenzaldehyde, 3-methoxypiperonal, 1,3-benzodioxole-5-carboxaldehyde, 7-methoxy, 5-methoxypiperonal, 7-methoxy-benzo 1,3 dioxole-5-carbaldehyde, 7-methoxybenzo d 1,3 dioxole-5-carbaldehyde, 7-methoxy-2h-1,3-benzodioxole-5-carbaldehyde, 7-methoxybenzo-1,3-dioxole-5-carboxaldehyde, 7-methoxybenzo 1,3 dioxole-5-carbaldehyde
InChI Key LOFRBHZYZCIOJO-UHFFFAOYSA-N
SMILES COC1=CC(=CC2=C1OCO2)C=O
IUPAC Name 7-methoxy-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol) 180.16
PubChem CID 22016
Formula Weight 246.35
Percent Purity ≥98%
Chemical Name or Material 1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.