accessibility menu, dialog, popup

UserName

HOOH

CAS RN 5768-10-5

IUPAC Name: 1-[4-(1-hydroxycyclohexyl)buta-1,3-diyn-1-yl]cyclohexan-1-ol
Synonyms: 1-[4-(1-hydroxycyclohexyl)buta-1,3-diyn-1-yl]cyclohexan-1-ol
Molecular Weight (g/mol): 246.35
Molecular Formula: C16H22O2
InChi Key: KJOFBMLTQUHDCL-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C#CC#CC1(O)CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results