Learn More
1,4-Benzenediboronic acid bis(neopentyl glycol) ester, 99%, Thermo Scientific™
Product Code. 11392917
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392917 1 g 1g
11312927 250 mg 250mg
11302927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11392917 Supplier Thermo Scientific Alfa Aesar Supplier No. L16187.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5565-36-6
Molecular Formula C16H24B2O4
Molecular Weight (g/mol) 301.984
MDL Number MFCD01075726
InChI Key XDODHEZHZCJSML-UHFFFAOYSA-N
Synonym 1,4-benzenediboronic acid bis neopentyl glycol ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinan-2-yl benzene, 2-4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl phenyl-5,5-dimethyl-1,3,2-dioxaborinane, 1,4-benzenediboronic acid bis npg cyclic ester, 1,4-benzenediboronic acid bisneopentylglycol cyclic ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinane-2-yl benzene, 1,4-benzenediboronic acid bis neopentyl glycol cyclic ester, 2,2'-1,4-phenylene bis 5,5-dimethyl-1,3,2-dioxaborinane
PubChem CID 23005407
IUPAC Name 2-[4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=C(C=C2)B3OCC(CO3)(C)C
Specifications
Melting Point 231°C to 233°C
Quantity 1 g
Beilstein 2952607
Formula Weight 301.99
Percent Purity 99%
Chemical Name or Material 1,4-Benzenediboronic acid bis(neopentyl glycol) ester