accessibility menu, dialog, popup

UserName

CAS RN 5565-36-6

IUPAC Name: 2-[4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-[4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 301.98
Molecular Formula: C16H24B2O4
InChi Key: XDODHEZHZCJSML-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=C(C=C1)B1OCC(C)(C)CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results