Learn More
1,3-Dichloro-5,5-dimethylhydantoin, TRC
Product Code. 30386111
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. Quantity
30386111 1 g
30372593 25 g
30388192 5 g
3 options
Product Code. 30386111
Brand: Toronto Research Chemicals TRCD4344301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118-52-5
Molecular Formula C5 H6 Cl2 N2 O2
Molecular Weight (g/mol) 197.02
Synonym 2,4-Imidazolidinedione, 1,3-dichloro-5,5-dimethyl-, N,N'-Dichlorodimethylhydantoin, 1,3-Dichloro-5,5-dimethylhydantoin, 1,3-Dichloro-5,5-dimethyl-imidazolidine-2,4-dione
IUPAC Name 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)N(Cl)C(=O)N(Cl)C1=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C5H6Cl2N2O2/c1-5(2)3(10)8(6)4(11)9(5)7/h1-2H3
Formula Weight 195.9806
Chemical Name or Material 1,3-Dichloro-5,5-dimethylhydantoin