accessibility menu, dialog, popup

UserName

CAS RN 118-52-5

CH3H3CNClONClO

CAS RN 118-52-5

IUPAC Name: 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
Synonyms: 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
Molecular Weight (g/mol): 197.02
Molecular Formula: C5H6Cl2N2O2
InChi Key: KEQGZUUPPQEDPF-UHFFFAOYSA-N
SMILES: CC1(C)N(Cl)C(=O)N(Cl)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results