Learn More
1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT
Product Code. 11359186
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359186 5 g 5g
11369186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11359186 Supplier Thermo Scientific Alfa Aesar Supplier No. L20334.06
Request Bulk or Custom Format
CAS: 926-57-8 | C4H6Cl2 | 124.99 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 926-57-8
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000861
InChI Key WLIADPFXSACYLS-DUXPYHPUSA-N
Synonym 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2
PubChem CID 643911
SMILES C\C(Cl)=C/CCl
Specifications
Density 1.161
Boiling Point 125°C to 129°C
Flash Point 33°C (91°F)
Refractive Index 1.473
Quantity 5 g
UN Number UN1992
Beilstein 906754
Sensitivity Moisture sensitive
IUPAC Name (2E)-1,3-dichlorobut-2-ene
Formula Weight 125
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-butene, Cis + Trans