accessibility menu, dialog, popup

UserName

CH3ClCl(E)

CAS RN 926-57-8

IUPAC Name: (2E)-1,3-dichlorobut-2-ene
Synonyms: 1,3-dichloro-2-butene, E-
Molecular Weight (g/mol): 124.99
Molecular Formula: C4H6Cl2
InChi Key: WLIADPFXSACYLS-DUXPYHPUSA-N
SMILES: C\C(Cl)=C/CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)