missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromoacetone, Technical Grade, TRC

Product Code. 30374022
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374022 50 g
30374760 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374022 Supplier Toronto Research Chemicals Supplier No. TRCD42277550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-39-7
Molecular Formula C3 H4 Br2 O
Molecular Weight (g/mol) 215.87
Synonym 2-Propanone, 1,3-dibromo-, 1,3-Dibromo-2-propanone, 1,3-Dibromoacetone, Bis(bromomethyl) ketone, NSC 249810
IUPAC Name 1,3-dibromopropan-2-one
SMILES BrCC(=O)CBr

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H4Br2O/c4-1-3(6)2-5/h1-2H2
Formula Weight 213.8629
Chemical Name or Material 1,3-Dibromoacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.