CAS RN 816-39-7
CAS RN 816-39-7
IUPAC Name:
1,3-dibromopropan-2-one
Synonyms:
2-propanone, 1,3-dibromo-
Molecular Weight (g/mol):
215.87
Molecular Formula:
C3H4Br2O
InChi Key:
LQQKDSXCDXHLLF-UHFFFAOYSA-N
SMILES:
BrCC(=O)CBr
1
–
3
of
3
results
1
1,3-Dibromoacetone, 95%
CAS: 816-39-7 Molecular Formula: C3H4Br2O Molecular Weight (g/mol): 215.872 MDL Number: MFCD00013540 InChI Key: LQQKDSXCDXHLLF-UHFFFAOYSA-N Synonym: 1,3-dibromoacetone,1,3-dibromo-2-propanone,2-propanone, 1,3-dibromo,1,3-dibromopropanone,bromomethyl ketone,bromomethylketone,sgqdllrxbrwsp@,1,3-dibromacetone,acmc-20ap4e,2-propanone,3-dibromo PubChem CID: 69952 IUPAC Name: 1,3-dibromopropan-2-one SMILES: C(C(=O)CBr)Br
3
1,3-Dibromoacetone, Technical Grade, TRC
CAS: 816-39-7 Molecular Formula: C3 H4 Br2 O Molecular Weight (g/mol): 215.87 Synonym: 2-Propanone, 1,3-dibromo-,1,3-Dibromo-2-propanone,1,3-Dibromoacetone,Bis(bromomethyl) ketone,NSC 249810 IUPAC Name: 1,3-dibromopropan-2-one SMILES: BrCC(=O)CBr