missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyladamantane, TRC

Product Code. 30457134
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457134 2.5 g 2.5g
30455209 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457134 Supplier Toronto Research Chemicals Supplier No. TRCT7957802.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-35-7
Molecular Formula C13 H22
Molecular Weight (g/mol) 178.31
Synonym Tricyclo[3.3.1.13,7]decane, 1,3,5-trimethyl-, Adamantane, 1,3,5-trimethyl- (7CI,8CI), 1,3,5-Trimethyltricyclo[3.3.1.13,7]decane, 1,3,5-Trimethyladamantane
IUPAC Name 1,3,5-trimethyladamantane
SMILES CC12CC3CC(C)(CC(C)(C3)C1)C2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H22/c1-11-4-10-5-12(2,7-11)9-13(3,6-10)8-11/h10H,4-9H2,1-3H3
Formula Weight 178.1722
Chemical Name or Material 1,3,5-Trimethyladamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.