CAS RN 707-35-7
CAS RN 707-35-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3,5-Trimethyladamantane, TRC
CAS: 707-35-7 Molecular Formula: C13 H22 Molecular Weight (g/mol): 178.31 Synonym: Tricyclo[3.3.1.13,7]decane, 1,3,5-trimethyl-,Adamantane, 1,3,5-trimethyl- (7CI,8CI),1,3,5-Trimethyltricyclo[3.3.1.13,7]decane,1,3,5-Trimethyladamantane IUPAC Name: 1,3,5-trimethyladamantane SMILES: CC12CC3CC(C)(CC(C)(C3)C1)C2