Learn More
1,2-Dichlorobenzene, Supelco™
Product Code. 15884310
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884310 2.5 L 2.5 L
15837150 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15884310 Supplier Supelco Supplier No. 1029302500
 Restricted Item
Estimated Shipment: 18-11-2026
For extraction analysis EMSURE™
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-50-1
Grade Research
Molecular Formula 1,2-(Cl)2C6H4
Molecular Weight (g/mol) 147
Specifications
Melting Point -17°C
Density 1.31 g/cm3 at 20°C
Quantity 2.5 L
MDL Number MFCD00000535
Synonym 1,2-Dichlorobenzene, o-Chlorobenzene, 1,2-Dichlorobenzene
Boiling Point 180°C/1013 hPa
Chemical Name or Material 1,2-Dichlorobenzene