CAS RN 95-50-1
1,2-Dichlorobenzene, 99%, pure
CAS: 95-50-1 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99+%, for HPLC
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 98+%, Extra Dry, AcroSeal™
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99+%, for spectroscopy
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, Extra Pure, SLR, Fisher Chemical™
CAS: 95-50-1 Molecular Formula: C6H4Cl2 MDL Number: 535
1,2-Dichlorobenzene, HPLC Grade, 98% min
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99%
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
Aromatic VOC Mixture 881 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
VOC Mixture 617 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Purgeable Aromatics Mixture 880 200 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
1,2-Dichlorobenzene, Supelco™
CAS: 95-50-1 Grade: Research Molecular Formula: 1,2-(Cl)2C6H4 Molecular Weight (g/mol): 147
1,2-Dichlorobenzene, Sigma-Aldrich™
CAS: 95-50-1 Grade: Research Molecular Formula: 1,2-(Cl)2C6H4 Molecular Weight (g/mol): 147
Purgeable Aromatic for Gas.Ident.Mix 3 200 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
- 1
- 2