missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,9-Tetrahydro-3-(1H-imidazol-1-ylmethyl)-9-methyl-4H-carbazol-4-one (Ondansetron Impurity G), TRC

Product Code. 30180368
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180368 10 mg 10mg
30179773 50 mg 50mg
30180026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180368 Supplier Toronto Research Chemicals Supplier No. TRCT29371510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99614-03-6
Molecular Formula C17 H17 N3 O
Molecular Weight (g/mol) 279.34
Synonym (3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, Ondansetron Hydrochloride Dihydrate Imp. G (EP), C-Demethylondansetron
IUPAC Name 3-(imidazol-1-ylmethyl)-9-methyl-2,3-dihydro-1H-carbazol-4-one
SMILES Cn1c2CCC(Cn3ccnc3)C(=O)c2c4ccccc14

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17N3O/c1-19-14-5-3-2-4-13(14)16-15(19)7-6-12(17(16)21)10-20-9-8-18-11-20/h2-5,8-9,11-12H,6-7,10H2,1H3
Formula Weight 279.1372
Percent Purity >95%
Chemical Name or Material C-Demethylondansetron
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.