CAS RN 99614-03-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,2,3,9-Tetrahydro-3-(1H-imidazol-1-ylmethyl)-9-methyl-4H-carbazol-4-one (Ondansetron Impurity G), TRC
CAS: 99614-03-6 Molecular Formula: C17 H17 N3 O Molecular Weight (g/mol): 279.34 Synonym: (3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one,Ondansetron Hydrochloride Dihydrate Imp. G (EP),C-Demethylondansetron IUPAC Name: 3-(imidazol-1-ylmethyl)-9-methyl-2,3-dihydro-1H-carbazol-4-one SMILES: Cn1c2CCC(Cn3ccnc3)C(=O)c2c4ccccc14
2
(3RS)-3-[(1H-Imidazol-1-yl)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one (C-Demethylondansetron), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.