missing translation for 'onlineSavingsMsg'
Learn More

1,2,10-Decanetriol, TRC

Product Code. 30365349
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365349 2 g 2g
30355471 10 g 10g
30349538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365349 Supplier Toronto Research Chemicals Supplier No. TRCD2125002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91717-85-0
Molecular Formula C10 H22 O3
Molecular Weight (g/mol) 190.28
IUPAC Name decane-1,2,10-triol
SMILES OCCCCCCCCC(O)CO

Specifications

Quantity 2 g
InChI Formula InChI=1S/C10H22O3/c11-8-6-4-2-1-3-5-7-10(13)9-12/h10-13H,1-9H2
Formula Weight 190.1569
Chemical Name or Material 1,2,10-Decanetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.