CAS RN 91717-85-0
CAS RN 91717-85-0
IUPAC Name:
decane-1,2,10-triol
Synonyms:
decane-1,2,10-triol
Molecular Weight (g/mol):
190.28
Molecular Formula:
C10H22O3
InChi Key:
RHINSRUDDXGHLV-UHFFFAOYNA-N
SMILES:
OCCCCCCCCC(O)CO
1
–
1
of
1
results
1
1,2,10-Decanetriol, TRC
CAS: 91717-85-0 Molecular Formula: C10 H22 O3 Molecular Weight (g/mol): 190.28 IUPAC Name: decane-1,2,10-triol SMILES: OCCCCCCCCC(O)CO