missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Dioxolan-2-ylmethyl)-N-methyl-4-(methylamino)-1H-imidazole-5-carboxamide, TRC

Product Code. 30135390
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135390 250 mg 250mg
30134303 100 mg 100mg
30012594 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135390 Supplier Toronto Research Chemicals Supplier No. TRCD487005250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1429636-74-7
Molecular Formula C10H16N4O3
Molecular Weight (g/mol) 240.26
Synonym Theophylline Impurity;
SMILES CNC(=O)c1c(NC)ncn1CC2OCCO2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H16N4O3/c1-11-9-8(10(15)12-2)14(6-13-9)5-7-16-3-4-17-7/h6-7,11H,3-5H2,1-2H3,(H,12,15)
Chemical Name or Material 1-(1,3-Dioxolan-2-ylmethyl)-N-methyl-4-(methylamino)-1H-imidazole-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.