CAS RN 1429636-74-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(1,3-Dioxolan-2-ylmethyl)-N-methyl-4-(methylamino)-1H-imidazole-5-carboxamide, TRC
CAS: 1429636-74-7 Molecular Formula: C10H16N4O3 Molecular Weight (g/mol): 240.26 Synonym: Theophylline Impurity; SMILES: CNC(=O)c1c(NC)ncn1CC2OCCO2