Learn More
1,1,1,3,8,10,10,10-Octachlorodecane, TRC
Product Code. 30424295
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424295 10 mg 10mg
30420203 5 mg -
30415254 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30424295 Supplier Toronto Research Chemicals Supplier No. TRCO18503510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 601523-23-3
Molecular Formula C10H14Cl8
Molecular Weight (g/mol) 417.84
Synonym Decane, 1,1,1,3,8,10,10,10-octachloro- (9CI, ACI), 1,1,1,3,8,10,10,10-Octachlorodecane (ACI)
IUPAC Name 1,1,1,3,8,10,10,10-octachlorodecane
SMILES ClC(CCCCC(Cl)CC(Cl)(Cl)Cl)CC(Cl)(Cl)Cl
Specifikationer
Quantity 10 mg
InChI Formula InChI=1S/C10H14Cl8/c11-7(5-9(13,14)15)3-1-2-4-8(12)6-10(16,17)18/h7-8H,1-6H2
Formula Weight 413.8604
Chemical Name or Material 1,1,1,3,8,10,10,10-Octachlorodecane
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.