CAS RN 601523-23-3
CAS RN 601523-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1,1,3,8,10,10,10-Octachlorodecane, TRC
CAS: 601523-23-3 Molecular Formula: C10H14Cl8 Molecular Weight (g/mol): 417.84 Synonym: Decane, 1,1,1,3,8,10,10,10-octachloro- (9CI, ACI),1,1,1,3,8,10,10,10-Octachlorodecane (ACI) IUPAC Name: 1,1,1,3,8,10,10,10-octachlorodecane SMILES: ClC(CCCCC(Cl)CC(Cl)(Cl)Cl)CC(Cl)(Cl)Cl