missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,10,11-Hexachloroundecane, TRC

Product Code. 30012475
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012475 5 mg 5mg
30010749 25 mg 25mg
30004255 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012475 Supplier Toronto Research Chemicals Supplier No. TRCH2947935MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601523-28-8
Molecular Formula C11 H18 Cl6
Molecular Weight (g/mol) 362.98
Synonym Undecane, 1,1,1,3,10,11-hexachloro- (9CI, ACI), 1,1,1,3,10,11-Hexachloroundecane (ACI)
IUPAC Name 1,1,1,3,10,11-hexachloroundecane
SMILES ClCC(Cl)CCCCCCC(Cl)CC(Cl)(Cl)Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H18Cl6/c12-8-10(14)6-4-2-1-3-5-9(13)7-11(15,16)17/h9-10H,1-8H2
Formula Weight 359.954
Chemical Name or Material 1,1,1,3,10,11-Hexachloroundecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.