CAS RN 601523-28-8
CAS RN 601523-28-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1,1,3,10,11-Hexachloroundecane, TRC
CAS: 601523-28-8 Molecular Formula: C11 H18 Cl6 Molecular Weight (g/mol): 362.98 Synonym: Undecane, 1,1,1,3,10,11-hexachloro- (9CI, ACI),1,1,1,3,10,11-Hexachloroundecane (ACI) IUPAC Name: 1,1,1,3,10,11-hexachloroundecane SMILES: ClCC(Cl)CCCCCCC(Cl)CC(Cl)(Cl)Cl