| Chemical Name or Material |
Aldosterone
|
| Name Note |
98%
|
| CAS |
52-39-1
|
| CAS Min % |
97.5
|
| CAS Max % |
100.0
|
| Color |
White
|
| Loss on Drying |
6.0% max. (105°C, 3 hrs)
|
| Infrared Spectrum |
Authentic
|
| Assay Percent Range |
98%
|
| Packaging |
Glass bottle
|
| Molecular Formula |
C21H28O5
|
| MDL Number |
MFCD00051136
|
| Quantity |
25 mg
|
| Beilstein |
08, IV, 3491
|
| Merck Index |
15, 218
|
| Specific Rotation Condition |
+ 155.00 (20.00°C c=0.1,CHCl3)
|
| Specific Rotation |
+150° to +162° (20°C, 589nm) (c=0.1, CHCl3)
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| Synonym |
aldosterone,electrocortin,aldocortin,aldocorten,aldocortene,+-aldosterone,elektrocortin,d-aldosterone,reichstein x,18-oxocorticosterone
|
| InChI Key |
PQSUYGKTWSAVDQ-ZVIOFETBSA-N
|
| SMILES |
CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4C(=O)CO)C=O)O
|
| IUPAC Name |
(8S,9S,10R,11S,13R,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-13-carbaldehyde
|
| Molecular Weight (g/mol) |
360.45
|
| PubChem CID |
5839
|
| ChEBI |
CHEBI:27584
|
| Formula Weight |
360.45
|
| Percent Purity |
≥97.5% (HPLC)
|
| Physical Form |
Crystalline Powder
|