accessibility menu, dialog, popup

UserName

CAS RN 99247-33-3

NaH3CNH3COOONHO

CAS RN 99247-33-3

IUPAC Name: sodium 4-(dipropylcarbamoyl)-4-(phenylformamido)butanoate
Synonyms: sodium 4-(dipropylcarbamoyl)-4-(phenylformamido)butanoate
Molecular Weight (g/mol): 356.4
Molecular Formula: C18H25N2NaO4
InChi Key: UFMWGCINVOIJSO-UHFFFAOYNA-M
SMILES: [Na+].CCCN(CCC)C(=O)C(CCC([O-])=O)NC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results