CAS RN 96192-34-6
CAS RN 96192-34-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Fructoselysine Dihydrochloride, TRC
CAS: 96192-34-6 Molecular Formula: C12H24N2O7 . 2(HCl) Molecular Weight (g/mol): 308.3323646 Synonym: (S)-1-[(5-Amino-1-carboxypentyl)amino]-1-deoxy-D-fructose Dihydrochloride,Fructoselysine Dihydrochloride,Fructosyllysine Dihydrochloride,Nα-(1-Deoxy-D-fructos-1-yl)-L-lysine Dihydrochloride; SMILES: OC1(CN[C@H](C(O)=O)CCCCN)[C@@H](O)[C@H](O)[C@H](O)CO1>>OC[C@@H]2[C@@H](O)[C@H](O)C(O)(CN[C@H](C(O)=O)CCCCN)O2