accessibility menu, dialog, popup

UserName

H2CN

CAS RN 96184-42-8

IUPAC Name: 4-(4-ethenylcyclohexyl)benzonitrile
Synonyms: 4-(4-ethenylcyclohexyl)benzonitrile
Molecular Weight (g/mol): 211.31
Molecular Formula: C15H17N
InChi Key: JHXJAWXZLWDDED-UHFFFAOYSA-N
SMILES: C=CC1CCC(CC1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results