accessibility menu, dialog, popup

UserName

CAS RN 96-70-8

H3COHH3CH3CCH3

CAS RN 96-70-8

IUPAC Name: 2-tert-butyl-4-ethylphenol
Synonyms: 2-(T-butyl)-4-ethylphenol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: LZHCVNIARUXHAL-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O)C(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results