Learn More
2-(tert-Butyl)-4-ethylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-tert-Butyl-4-ethylphenol |
| CAS | 96-70-8 |
| Molecular Formula | C12 H18 O |
| Quantity | 5 g |
| Synonym | 2-(1,1-Dimethylethyl)-4-ethylphenol, 2-tert-Butyl-4-ethylphenol, 4-Ethyl-2-tert-butylphenol |
| InChI Formula | InChI=1S/C12H18O/c1-5-9-6-7-11(13)10(8-9)12(2,3)4/h6-8,13H,5H2,1-4H3 |
| SMILES | CCc1ccc(O)c(c1)C(C)(C)C |
| IUPAC Name | 2-tert-butyl-4-ethylphenol |
| Molecular Weight (g/mol) | 178.27 |
| Formula Weight | 178.1358 |