CAS RN 954230-39-8
IUPAC Name:
ethyl 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylate
Synonyms:
ethyl 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol):
249.24
Molecular Formula:
C13H12FNO3
InChi Key:
QQZTUBJEZZHCKF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)ON=C1C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Ethyl 3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate, 97%, Thermo Scientific™
CAS: 954230-39-8 Molecular Formula: C13H12FNO3 Molecular Weight (g/mol): 249.24 MDL Number: MFCD09756447 InChI Key: QQZTUBJEZZHCKF-UHFFFAOYSA-N Synonym: ethyl 3-4-fluorophenyl-5-methylisoxazole-4-carboxylate,3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester,3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid ethylester,ethyl 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylate,3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid ethyl ester,4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl-, ethyl PubChem CID: 22309061 IUPAC Name: ethyl 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylate SMILES: CCOC(=O)C1=C(C)ON=C1C1=CC=C(F)C=C1