accessibility menu, dialog, popup

UserName

CAS RN 95333-95-2

IUPAC Name: 3-(3,5-dichlorophenyl)propanoic acid
Synonyms: 3-(3,5-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: GQICJNXGCGRFBA-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results