accessibility menu, dialog, popup

UserName

CAS RN 950603-19-7

NNOOH

CAS RN 950603-19-7

IUPAC Name: 5-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Synonyms: 5-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: ACLFTMQROMOTLA-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results