CAS RN 945-32-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyrrole-2,3,5-tricarboxylic Acid, TRC
CAS: 945-32-4 Molecular Formula: C7H5NO6 Molecular Weight (g/mol): 199.12 Synonym: 1H-Pyrrole-2,3,5-tricarboxylic Acid IUPAC Name: 1H-pyrrole-2,3,5-tricarboxylic acid SMILES: C1=C(C(=O)O)NC(=C1C(=O)O)C(=O)O