accessibility menu, dialog, popup

UserName

H3CH3COHONON

CAS RN 94470-67-4

IUPAC Name: 3-hydroxy-2,2-dimethyl-4-(2-oxopyrrolidin-1-yl)-3,4-dihydro-2H-1-benzopyran-6-carbonitrile
Synonyms: cromakalim (3s-trans)-cromakalim (3s-trans)-isomer cromakalim cromakalim, (3R-trans)-isomer (3S-cis)-isomer cromakalim cromakalim, (3R-cis)- cromakalimum cromakalim, (3R-trans)- cromakalim, (3S-trans)- (3S-trans)-cromakalim
Molecular Weight (g/mol): 286.33
Molecular Formula: C16H18N2O3
InChi Key: TVZCRIROJQEVOT-UHFFFAOYNA-N
SMILES: CC1(C)OC2=CC=C(C=C2C(C1O)N1CCCC1=O)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)