Learn More
(+/-)-Cromakalim, TRC
Specifications
Specifications
| Chemical Name or Material | (+/-)-Cromakalim |
| CAS | 94470-67-4 |
| Molecular Formula | C16 H18 N2 O3 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C16H18N2O3/c1-16(2)15(20)14(18-7-3-4-13(18)19)11-8-10(9-17)5-6-12(11)21-16/h5-6,8,14-15,20H,3-4,7H2,1-2H3/t14-,15+/m1/s1 |
| SMILES | CC1(C)Oc2ccc(cc2[C@H]([C@@H]1O)N3CCCC3=O)C#N |
| IUPAC Name | (3S,4R)-3-hydroxy-2,2-dimethyl-4-(2-oxopyrrolidin-1-yl)-3,4-dihydrochromene-6-carbonitrile |
| Molecular Weight (g/mol) | 286.33 |
| Formula Weight | 286.1317 |