accessibility menu, dialog, popup

UserName

CAS RN 942206-12-4

NIHNNH2Cl

CAS RN 942206-12-4

IUPAC Name: 5-chloro-2-hydrazinyl-3-iodopyridine
Synonyms: 5-chloro-2-hydrazinyl-3-iodopyridine
Molecular Weight (g/mol): 269.47
Molecular Formula: C5H5ClIN3
InChi Key: SMVJKHCZAOIBLS-UHFFFAOYSA-N
SMILES: NNC1=C(I)C=C(Cl)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results