accessibility menu, dialog, popup

UserName

OHNHONH2(E/Z)

CAS RN 939999-37-8

IUPAC Name: N'-hydroxy-3-(hydroxymethyl)benzene-1-carboximidamide
Synonyms: N'-hydroxy-3-(hydroxymethyl)benzenecarboximidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: DKSFMGRTWCDFRB-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(CO)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results