accessibility menu, dialog, popup

UserName

H3CH2NOOCH3

CAS RN 93750-30-2

IUPAC Name: 2-(4-methoxyphenoxy)propan-1-amine
Synonyms: 2-(4-methoxyphenoxy)propan-1-amine
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: HIWWBKQBOHWJIM-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC(C)CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results