CAS RN 932986-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Azoramide, MedChemExpress
MedChemExpress Azoramide is a small-molecule modulator of the unfolded protein response with antidiabetic activity.