Learn More
Azoramide, MedChemExpress
Description
Azoramide is a small-molecule modulator of the unfolded protein response with antidiabetic activity.
Specifications
Specifications
| Chemical Name or Material | Azoramide |
| Physical Form | Solid |
| CAS | 932986-18-0 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H17ClN2OS |
| Solubility Information | DMSO : ≥ 30 mg/mL (97.14 mM) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) |
| SMILES | CCCC(NCCC1=CSC(C2=CC=C(Cl)C=C2)=N1)=O |
| Molecular Weight (g/mol) | 308.83 |
| Show More |