CAS RN 928-94-9
CAS RN 928-94-9
IUPAC Name:
(2Z)-hex-2-en-1-ol
Synonyms:
(2Z)-hex-2-en-1-ol
Molecular Weight (g/mol):
100.16
Molecular Formula:
C6H12O
InChi Key:
ZCHHRLHTBGRGOT-PLNGDYQASA-N
SMILES:
CCC\C=C/CO
1
–
3
of
3
results
1
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
CAS: 928-94-9 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00063209 InChI Key: ZCHHRLHTBGRGOT-PLNGDYQASA-N Synonym: cis-2-hexen-1-ol,2-hexen-1-ol, z,cis-hex-2-en-1-ol,z-2-hexen-1-ol,z-hex-2-en-1-ol,unii-871tel510e,2z-hexen-1-ol,2z-hex-2-en-1-ol,cis-1-hydroxy-2-hexene,cis-2-hexenol PubChem CID: 5324489 IUPAC Name: (Z)-hex-2-en-1-ol SMILES: CCC\C=C/CO
2
cis-2-Hexen-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-2-Hexen-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.