Learn More
cis-2-Hexen-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | cis-2-Hexen-1-ol |
| CAS | 928-94-9 |
| Molecular Formula | C6H12O |
| Quantity | 10 g |
| Synonym | (2Z)-2-Hexen-1-ol, (Z)-1-Hydroxy-2-hexene, (Z)-2-Hexen-1-ol, (Z)-2-Hexenol, cis-2-Hexenol, cis-beta,gamma-Hexenol |
| InChI Formula | InChI=1S/C6H12O/c1-2-3-4-5-6-7/h4-5,7H,2-3,6H2,1H3/b5-4- |
| SMILES | CCC\C=C/CO |
| IUPAC Name | (Z)-hex-2-en-1-ol |
| Molecular Weight (g/mol) | 100.16 |
| Formula Weight | 100.0888 |