accessibility menu, dialog, popup

UserName

CH3OHNI

CAS RN 927870-76-6

IUPAC Name: 5-iodo-6-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-iodo-6-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 235.02
Molecular Formula: C6H6INO
InChi Key: UFZSHCNPDKACCS-UHFFFAOYSA-N
SMILES: CC1=C(I)C=CC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results